Learn More
Thermo Scientific NanoViper Capillary 20um ID x 750mm, New (1)
Supplier:  Thermo Scientific 6041.5280
Encompass
fused silica capillary tubing for chromatography with inner diameter 20 micrometers, outer diameter 320 micrometers, polyimide external coating, flexible, high mechanical strength, chemically inert, for high performance liquid chromatography applications.
Catalog No. 50-134-7910