Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier:  Thermo Scientific 6041.5560
Encompass
- Virtually zero-dead-volume connections ensure optimal separation efficiencies- Preassembled and finger-tightened for easy tool-free instrument setup- High-pressure tested for UHPLC compatibility up to 1200 bar (17400 psi)- Suitable for temperatures up to 120C- Compatible with valves and columns from virtually any manufacturer
Catalog No. 50-134-7921