Learn More
Thermo Scientific Viper Fingertight Fitting Systems | PEEK | 1 EA
Catalog No. 501347925 Shop All Thermo Scientific Products
Encompass
Ferrule-free tightening design for virtually zero-dead-volume connections, Operates at pressures up to 345 bar, Fingertight design, Flexible biocompatible PEEK capillaries 1/32 in OD, No tools required, Compatible with virtually any manufacturer valve and column, Fits narrow connections such as 10-port valves, Available in different materials lengths and inner diameters
Catalog No. 50-134-7925 Supplier Thermo Scientific Supplier No. 6041.5615
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ferrule-free tightening design for virtually zero-dead-volume connections, Operates at pressures up to 345 bar, Fingertight design, Flexible biocompatible PEEK capillaries 1/32 in OD, No tools required, Compatible with virtually any manufacturer valve and column, Fits narrow connections such as 10-port valves, Available in different materials lengths and inner diameters