Learn More
Thermo Scientific NANOVIPER CAPILLARY IDXL 20UM
Supplier:  Thermo Scientific 6041.5240
Encompass
A narrow bore 20 micrometer internal diameter capillary tube for liquid sample injection and microfluidic sample transport enabling nanoliter sample volume handling for analytical instrumentation.
Catalog No. NC1627463