Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier:  Thermo Scientific 6041.5828
Encompass
- Virtually zero-dead-volume connections ensure optimal separation efficiencies- Preassembled and finger-tightened for easy tool-free setup- High-pressure tested, compatible up to 1200 bar (17400 psi)- Suitable for temperatures up to 120C
Catalog No. NC1372308