Learn More
Beckman Coulter nanoViS Nanoscale Sizing Standards Kit - polystyrene bead calibration set for nanoscale flow cytometry (Beckman Coulter, PN D03231)
Catalog No. NC3804425
Encompass
The nanoViS Nanoscale Sizing Standards Kit (PN D03231) is a set of polystyrene reference beads for calibrating and verifying light-scatter-based nanoscale flow cytometers. The kit provides defined low- and high-scatter bead populations to generate scatter-to-size calibration curves and support routine instrument performance checks.
  • Includes low-scatter polystyrene beads: 40 nm, 80 nm, 108 nm, 142 nm.
  • Includes high-scatter polystyrene beads: 142 nm, 304 nm, 600 nm, 1020 nm.
  • One vial provided for each nominal bead size.
  • Enables scatter-to-size calibration using Mie theory.
  • Intended for instrument calibration and routine performance verification in nanoscale flow cytometry.
Catalog No. NC3804425 Supplier Beckman Coulter Supplier No. D03231
The nanoViS Nanoscale Sizing Standards Kit (PN D03231) is a set of polystyrene reference beads for calibrating and verifying light-scatter-based nanoscale flow cytometers. The kit provides defined low- and high-scatter bead populations to generate scatter-to-size calibration curves and support routine instrument performance checks.
  • Includes low-scatter polystyrene beads: 40 nm, 80 nm, 108 nm, 142 nm.
  • Includes high-scatter polystyrene beads: 142 nm, 304 nm, 600 nm, 1020 nm.
  • One vial provided for each nominal bead size.
  • Enables scatter-to-size calibration using Mie theory.
  • Intended for instrument calibration and routine performance verification in nanoscale flow cytometry.