Learn More
Nanocomposix NanoXact Silver Nanospheres - Bare (Citrate) - 50 nm, 0.02 mg/mL in 2 mM sodium citrate, 50 mL
SDP
Catalog No. NC2174043
Encompass_Preferred
As a global leader in silver nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.50 nm spherical silver nanoparticles combine strong plasmonic optical properties with antimicrobial capabilities. These nanoparticles are useful in a broad range of applications including conductive composites, biosensors, sensing, photovoltaics, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, and UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC2174043 Supplier Nanocomposix Supplier No. AGCN5050M
As a global leader in silver nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.50 nm spherical silver nanoparticles combine strong plasmonic optical properties with antimicrobial capabilities. These nanoparticles are useful in a broad range of applications including conductive composites, biosensors, sensing, photovoltaics, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, and UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.