Learn More
Aviva Systems Biology Corp NANP Antibody - middle region (ARP53100_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53100_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NANP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NANP. Alternate names: C20orf147, HDHD4, MGC26833, dJ694B14.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60920