Learn More
Aviva Systems Biology Corp NANS Antibody - C-terminal region (ARP65353_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NANS. It was validated on Western Blot. . Alternate names: SASApplicationWB. Species reactivity: Cow, Dog, Human, Pig, Rabbit, Rat
Catalog No. 50-288-483