Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMarKa Chemical Company Sodium Hydroxide 0.01N(in Methanol), 1l

Catalog No. 504400358
Encompass_Preferred

Sodium Hydroxide 0.01N(in Methanol), 1l Plastic Bottle

Catalog No. 50-440-0358 Supplier LaMarKa Chemical Company Supplier No. 0000155816750101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Hydroxide 0.01N(in Methanol), 1l Plastic Bottle

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.