Learn More
Medchemexpress LLC NAP-2/CXCL7 Human 10ug
SDP
Catalog No. 5000182234
Encompass_Preferred
CXCL7 (also known as neutrophil activating peptide 2 NAP-2) is a platelet-derived growth factor that belongs to the ELR CXC chemokine family functioning as a potent chemoattractant and activator of neutrophils through binding to its receptor CXCR2[1] NAP-2/CXCL7 Protein Human (HEK 293 His) is produced in HEK293 cells with six C-Terminal His-tags It consists of 94 amino acids (S35-D128)
Catalog No. 50-001-82234 Supplier Medchemexpress LLC Supplier No. HYP7267610UG
CXCL7 (also known as neutrophil activating peptide 2 NAP-2) is a platelet-derived growth factor that belongs to the ELR CXC chemokine family functioning as a potent chemoattractant and activator of neutrophils through binding to its receptor CXCR2[1] NAP-2/CXCL7 Protein Human (HEK 293 His) is produced in HEK293 cells with six C-Terminal His-tags It consists of 94 amino acids (S35-D128)