Learn More
Medchemexpress LLC NAP-2/CXCL7 Human 10ug
SDP
Catalog No. 5000182857
Encompass_Preferred
CXCL7 (also known as neutrophil activating peptide 2 NAP-2) is a platelet-derived growth factor that belongs to the ELR CXC chemokine family functioning as a potent chemoattractant and activator of neutrophils through binding to its receptor CXCR2[1] NAP-2/CXCL7 Protein Human (CHO) is produced in CHO cells and consists of 70 amino acids (A59-D128)
Catalog No. 50-001-82857 Supplier Medchemexpress LLC Supplier No. HYP726710UG
CXCL7 (also known as neutrophil activating peptide 2 NAP-2) is a platelet-derived growth factor that belongs to the ELR CXC chemokine family functioning as a potent chemoattractant and activator of neutrophils through binding to its receptor CXCR2[1] NAP-2/CXCL7 Protein Human (CHO) is produced in CHO cells and consists of 70 amino acids (A59-D128)