Learn More
MOLTOX Molecular Toxicology Inc Nutrient Agar, Oxoid #2, 30 mL/plate, 20 plates/sleeve, Store at Room Temperature
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 2110S.1
Encompass
Nutrient Agar is a general purpose medium for the cultivation and maintenance of nonfastidious organisms. It can be used for the cultivation and enumeration of organisms in water, sewage, feces and other materials. Nutrient agars prepared with Oxoid Nutrient Broth No. 2, specified by Maron & Ames, Mutat. Res. 113: 173, 1983, are used for general growth and CFU determinations of Ames S. typhimurium and E. coli WP2 strains. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-607