Learn More
Carbosynth LLC. NAP Peptide, 25mg, CAS# 11439-12-2, MF= C36H60N10O12
SDP
Supplier:  Carbosynth LLC. NAP3012PI25MG
Encompass_Preferred
Davunetide; ADNP Neuropeptide Fragment; H-Asn-Ala-Pro-Val-Ser-Ile-Pro-Gln-OH, Application: Short Neuropeptide Fragment Derived from Activity-Dependent Neuroprotective Protein (ADNP); May Have Neuroprotective Effects Against Schizophrenia-Related Impairment, Toxicity Associated with Beta-Amyloid and Other Compounds, Notes: NAP peptide is a short neuropeptide fragment derived from Activity-Dependent Neuroprotective Protein (ADNP). This octopeptide may have neuroprotective effects against schizophrenia-related impairment, toxicity associated with beta-amyloid and other compounds, stabilizing microtuble breakdown which may suggest therapeutic strategies for Parkinson's, Alzheimer's and related diseases., salt: Trifluoroacatate Salt, one letter code: NAPVSIPQ, MW: 824.94
Catalog No. 50-195-2413