Learn More
Bioworld NAP- (Non-Animal Protein) Blocker, 1 L
Supplier:  Bioworld 209100001
Encompass_Preferred
NAP (Non Animal Protein)-BLOCKER provides improved assay sensitivity, clean background and high signal-to-background ratio. Produced from non-animal proteins that are free of biotin and other agents that interfere with immunoassays. Will not cross react with animal source antigens or primary and secondary antibodies. Supplied as a 4X solution, simply mix with water to obtain a 1X ready to use buffer for nitrocellulose and PVDF membranes. Suitable for up to 100 mini-gel blots using plastic bags for reduced volume. Usually Ships within 24 HoursAdvantage:Reduces risk of non-specific binding Lowers background Improves assay sensitivityApplication: Western blot, dot blot & ELISA experiments
Catalog No. 50-198-522