Learn More
Aviva Systems Biology Corp NAP1L2 Antibody - middle region (ARP57574_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57574_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NAP1L2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human NAP1L2. Alternate names: BPX, MGC26243ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-335