Learn More
Aviva Systems Biology Corp NAPA Antibody - N-terminal region (AVARP13049_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13049_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NAPA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NAPA. Alternate names: SNAPAApplicationWB. Species reactivity: Cow, Dog, Mouse, Zebrafish
Catalog No. 50-014-53790