Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter™ NAPA Calibrator Emit 2000

Catalog No. 01801135 Shop All Beckman Coulter Products
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-801-135 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-801-135 Supplier Beckman Coulter™ Supplier No. 4N109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NAPA Calibrator for use with Beckman Coulter analyzers.

  • For use with Beckman Coulter analyzers.
  • Includes 1 x 5 mL and 5 x 2 mL
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Beckman Coulter Analyzers
Quantity 15 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.