Learn More
Se Lab Group Inc NAPA Nectar 4oz, 50pch/Case
SDP
Catalog No. NC9479557
Encompass
The most widely used sterile water gel for laboratory animal hydration. Ideal for: • Shipping • Disaster planning • Animal Quarantine • Biocontainment suites. • USDA approved ingredients• Moisture content 99.35% by volume • Non-refrigerated shelf life of 24 months • Packaged in a disposable inert poly pouch
Catalog No. NC9479557 Supplier Se Lab Group Inc Supplier No. 880004
The most widely used sterile water gel for laboratory animal hydration. Ideal for: • Shipping • Disaster planning • Animal Quarantine • Biocontainment suites. • USDA approved ingredients• Moisture content 99.35% by volume • Non-refrigerated shelf life of 24 months • Packaged in a disposable inert poly pouch