Learn More
Medchemexpress LLC HY-13919 10mg , Napabucasin CAS:83280-65-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1391910MG
Encompass_Preferred
HY-13919 10mg Napabucasin CAS: 83280-65-3 Napabucasin (BBI608) is a STAT3 inhibitor which blocks stem cell activity in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0823