Learn More
Medchemexpress LLC HY-13919 5mg Medchemexpress, Napabucasin CAS:83280-65-3 Purity:>98%
SDP
Catalog No. 502030492
Encompass_Preferred
Medchemexpress, HY-13919 5mg Napabucasin CAS:83280-65-3 Napabucasin is a STAT3 inhibitor which blocks stem cell activity in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0492 Supplier Medchemexpress LLC Supplier No. HY139195MG
Medchemexpress, HY-13919 5mg Napabucasin CAS:83280-65-3 Napabucasin is a STAT3 inhibitor which blocks stem cell activity in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.