Learn More
Selleck Chemical LLC Napabucasin5mg
SDP
Supplier:  Selleck Chemical LLC S79775MG
Encompass_Preferred
Napabucasin (BBI608) is an orally available Stat3 and cancer cell stemness inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3333