Learn More
Cayman Chemical NaphazolIn-d4hydrochlorIde 1mg
SDP
Catalog No. 502948360
Encompass_Preferred
An internal standard for the quantification of naphazoline by GC- or LC-MS
Catalog No. 50-294-8360 Supplier Cayman Chemical Supplier No. 340121 MG
An internal standard for the quantification of naphazoline by GC- or LC-MS