Learn More
Cayman Chemical NaphazolIn-d4hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 340125 MG
Encompass_Preferred
An internal standard for the quantification of naphazoline by GC- or LC-MS
Catalog No. 50-294-7485