Learn More
Medchemexpress LLC Naphazoline | 835-31-4 | 99.7% | 210.27 | 100 MG
SDP
Catalog No. 5000346462
Encompass_Preferred
Naphazoline (Naphthazoline) is a potent α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It can also reduce levels of inflammatory factors, cytokines, IgE, GMCSF, and NGF, making it suitable for non-bacterial conjunctivitis research.
  • Reduces vascular hyperpermeability
  • Promotes vasoconstriction
  • Reduces levels of inflammatory factors (TNF-α, IL-1β and IL-6)
  • Reduces levels of cytokines (IFN-γ and IL-4)
  • Reduces levels of IgE, GMCSF, and NGF
  • Can be used for non-bacterial conjunctivitis research
Catalog No. 50-003-46462 Supplier Medchemexpress LLC Supplier No. HY11132650MG
Naphazoline (Naphthazoline) is a potent α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It can also reduce levels of inflammatory factors, cytokines, IgE, GMCSF, and NGF, making it suitable for non-bacterial conjunctivitis research.
  • Reduces vascular hyperpermeability
  • Promotes vasoconstriction
  • Reduces levels of inflammatory factors (TNF-α, IL-1β and IL-6)
  • Reduces levels of cytokines (IFN-γ and IL-4)
  • Reduces levels of IgE, GMCSF, and NGF
  • Can be used for non-bacterial conjunctivitis research