Learn More
Medchemexpress LLC Naphazoline | 835-31-4 | MFCD00066737 | 99.7% | 210.27 | 25 MG
SDP
Catalog No. 5000346149
Encompass_Preferred
Naphazoline is a potent α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It also reduces the levels of inflammatory factors (TNF-α, IL-1β, and IL-6), cytokines (IFN-γ and IL-4), IgE, GMCSF, and NGF. It can be used for non-bacterial conjunctivitis research.
  • Reduces vascular hyperpermeability
  • Promotes vasoconstriction
  • Reduces inflammatory factors and cytokines
  • Used for non-bacterial conjunctivitis research
Catalog No. 50-003-46149 Supplier Medchemexpress LLC Supplier No. 25MGHY111326
Naphazoline is a potent α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It also reduces the levels of inflammatory factors (TNF-α, IL-1β, and IL-6), cytokines (IFN-γ and IL-4), IgE, GMCSF, and NGF. It can be used for non-bacterial conjunctivitis research.
  • Reduces vascular hyperpermeability
  • Promotes vasoconstriction
  • Reduces inflammatory factors and cytokines
  • Used for non-bacterial conjunctivitis research