Learn More
Medchemexpress LLC Naphazoline hydrochloride | 550-99-2 | 99.4% | 5 G
SDP
Catalog No. 5000406325
Encompass_Preferred
Naphazoline hydrochloride is a potent α-adrenergic receptor agonist. It reduces vascular hyperpermeability and promotes vasoconstriction. This compound also decreases the levels of inflammatory factors (TNF-α, IL-1β, IL-6), cytokines (IFN-γ, IL-4), IgE, GMCSF, and NGF. It is used in research for non-bacterial conjunctivitis.
  • Reduces vascular hyperpermeability
  • Promotes vasoconstriction
  • Decreases inflammatory factors (TNF-α, IL-1β, IL-6)
  • Decreases cytokines (IFN-γ, IL-4)
  • Decreases IgE, GMCSF, and NGF levels
  • Used in research for non-bacterial conjunctivitis
Catalog No. 50-004-06325 Supplier Medchemexpress LLC Supplier No. HYB04465G
Naphazoline hydrochloride is a potent α-adrenergic receptor agonist. It reduces vascular hyperpermeability and promotes vasoconstriction. This compound also decreases the levels of inflammatory factors (TNF-α, IL-1β, IL-6), cytokines (IFN-γ, IL-4), IgE, GMCSF, and NGF. It is used in research for non-bacterial conjunctivitis.
  • Reduces vascular hyperpermeability
  • Promotes vasoconstriction
  • Decreases inflammatory factors (TNF-α, IL-1β, IL-6)
  • Decreases cytokines (IFN-γ, IL-4)
  • Decreases IgE, GMCSF, and NGF levels
  • Used in research for non-bacterial conjunctivitis