Learn More
Medchemexpress LLC Naphazoline nitrate | 5144-52-5 | 99.1% | 273.29 | 100 MG
SDP
Catalog No. 5000346341
Encompass_Preferred
Naphazoline nitrate is an α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It also reduces levels of inflammatory factors and can be used for non-bacterial conjunctivitis research.
  • Targets α-adrenergic receptors
  • Helps manage vascular issues
  • Decreases levels of TNF-α, IL-1β, IL-6, IFN-γ, IL-4, IgE, GMCSF, and NGF
  • Applicable for non-bacterial conjunctivitis research
  • Solid, white to off-white appearance
  • Solubility: 100 mg/mL in DMSO
Catalog No. 50-003-46341 Supplier Medchemexpress LLC Supplier No. HY111326A5G
Naphazoline nitrate is an α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It also reduces levels of inflammatory factors and can be used for non-bacterial conjunctivitis research.
  • Targets α-adrenergic receptors
  • Helps manage vascular issues
  • Decreases levels of TNF-α, IL-1β, IL-6, IFN-γ, IL-4, IgE, GMCSF, and NGF
  • Applicable for non-bacterial conjunctivitis research
  • Solid, white to off-white appearance
  • Solubility: 100 mg/mL in DMSO