Learn More
Medchemexpress LLC Naphazoline nitrate | 5144-52-5 | MFCD00014316 | 99.1% | 273.29 | 50 MG
SDP
Catalog No. 5000346340
Encompass_Preferred
Naphazoline nitrate is an α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It is also known to reduce levels of inflammatory factors, cytokines, IgE, GMCSF, and NGF, making it suitable for non-bacterial conjunctivitis research.
  • α-adrenergic receptor agonist
  • Reduces vascular hyperpermeability
  • Promotes vasoconstriction
  • Reduces levels of inflammatory factors (TNF-α, IL-1β, IL-6)
  • Reduces levels of cytokines (IFN-γ, IL-4)
  • Reduces IgE, GMCSF, and NGF
  • Can be used for non-bacterial conjunctivitis research
Catalog No. 50-003-46340 Supplier Medchemexpress LLC Supplier No. HY111326A1G
Naphazoline nitrate is an α-adrenergic receptor agonist that reduces vascular hyperpermeability and promotes vasoconstriction. It is also known to reduce levels of inflammatory factors, cytokines, IgE, GMCSF, and NGF, making it suitable for non-bacterial conjunctivitis research.
  • α-adrenergic receptor agonist
  • Reduces vascular hyperpermeability
  • Promotes vasoconstriction
  • Reduces levels of inflammatory factors (TNF-α, IL-1β, IL-6)
  • Reduces levels of cytokines (IFN-γ, IL-4)
  • Reduces IgE, GMCSF, and NGF
  • Can be used for non-bacterial conjunctivitis research