Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphtha, VM and P, Spectrum™ Chemical
SDP

Catalog No. 18604745 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
4 L
20 L
4 x 4 L
200 L
4 x 20 L
Packaging:
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-745 200 L Steel Can
18-604-743 4 L Steel Can
18-604-744 4 x 4 L Steel Can
18-604-741 20 L Steel Can
18-604-742 4 x 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-745 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N1002200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N1002, 8030-30-6

Spectrum™ Chemical Naphtha, VM and P

Chemical Identifiers

CAS 8030-30-6

Specifications

CAS Max % 1
Quantity 200 L
Packaging Steel Can
UN Number UN1268
Boiling Point 118°C to 150°C
Grade Reagent
Chemical Name or Material Naphtha
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.