Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthacene 97.0+%, TCI America™
SDP

Catalog No. N00011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N00011G 1 g
N0001100MG 100 mg
N00015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N00011G Supplier TCI America Supplier No. N00011G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 92-24-0
Molecular Formula C18H12
Molecular Weight (g/mol) 228.294
MDL Number MFCD00003702
InChI Key IFLREYGFSNHWGE-UHFFFAOYSA-N
Synonym naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
PubChem CID 7080
ChEBI CHEBI:32600
IUPAC Name tetracene
SMILES C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1

Specifications

Melting Point 357°C
Quantity 1 g
Formula Weight 228.29
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Naphthacene
RTECSNumber QI7605000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.