Learn More
Medchemexpress LLC Naphthalene-2,3-dicarboxaldehyde | 7149-49-7 | MFCD00016618 | 99.3% | 184.19 g/mol | C12H8O2 | 50 MG
SDP
Catalog No. 5000302229
Encompass_Preferred
Naphthalene-2,3-dicarboxaldehyde is an aromatic dialdehyde used as a biochemical reagent and derivatization agent for primary amines. It is employed in fluorescence labeling, enzyme inhibition studies, and other research applications where a high-purity aromatic aldehyde is required.
  • High purity suitable for analytical and research use.
  • Reacts with primary amines to form highly fluorescent derivatives.
  • Reported enzyme inhibition activity useful for biochemical assays.
  • Available in small milligram-scale quantities for laboratory experiments.
  • Stable solid form with defined molecular weight and formula for reproducible results.
Catalog No. 50-003-02229 Supplier Medchemexpress LLC Supplier No. HYW01628850MG
Naphthalene-2,3-dicarboxaldehyde is an aromatic dialdehyde used as a biochemical reagent and derivatization agent for primary amines. It is employed in fluorescence labeling, enzyme inhibition studies, and other research applications where a high-purity aromatic aldehyde is required.
  • High purity suitable for analytical and research use.
  • Reacts with primary amines to form highly fluorescent derivatives.
  • Reported enzyme inhibition activity useful for biochemical assays.
  • Available in small milligram-scale quantities for laboratory experiments.
  • Stable solid form with defined molecular weight and formula for reproducible results.