Learn More
Medchemexpress LLC Naphthalene-2,3-dicarboxaldehyde | 7149-49-7 | MFCD00016618 | 99.4% | 184.19 | C12H8O2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW016288500MG
Encompass_Preferred
Naphthalene-2,3-dicarboxaldehyde is an aromatic dialdehyde used as a reagent in analytical and biochemical research. It is commonly used for fluorogenic derivatization of primary amines and for sensitive detection in assays. The compound is supplied as a high-purity solid with supporting documentation for research use.
  • High purity suitable for analytical applications.
  • Fluorogenic derivatization reagent for primary amines.
  • Supplied as a stable solid for convenient handling and storage.
  • Available in multiple pack sizes to support varied research needs.
  • Accompanied by datasheet and certificate of analysis documentation.
Catalog No. 50-003-03251