Learn More
Medchemexpress LLC Naphthalene-2,3-dicarboxaldehyde | 7149-49-7 | MFCD00016618 | 99.4% | 184.19 g/mol | C12H8O2 | 100 MG
SDP
Catalog No. 5000303694
Encompass_Preferred
Naphthalene-2,3-dicarboxaldehyde is a small organic aldehyde used as a research reagent for derivatization and biochemical assays. It is supplied as a solid with high reported purity and has documented activity in fungal enzyme inhibition and antifungal assays.
  • High purity (99.4%) suitable for research use.
  • Useful derivatization reagent for primary amines, amino acids, and small peptides.
  • Documented inhibitory activity against fungal ASADH (Ki ≈45 μM) and C. albicans (IC50 58.2 μM).
  • Available in multiple pack sizes including 100 mg for lab-scale experiments.
  • Solid, off-white to pink appearance for easy handling and storage.
Catalog No. 50-003-03694 Supplier Medchemexpress LLC Supplier No. HYW016288100MG
Add to Cart
Add to Cart
Naphthalene-2,3-dicarboxaldehyde is a small organic aldehyde used as a research reagent for derivatization and biochemical assays. It is supplied as a solid with high reported purity and has documented activity in fungal enzyme inhibition and antifungal assays.
  • High purity (99.4%) suitable for research use.
  • Useful derivatization reagent for primary amines, amino acids, and small peptides.
  • Documented inhibitory activity against fungal ASADH (Ki ≈45 μM) and C. albicans (IC50 58.2 μM).
  • Available in multiple pack sizes including 100 mg for lab-scale experiments.
  • Solid, off-white to pink appearance for easy handling and storage.