Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naphthalene-2,3-dicarboxaldehyde | 7149-49-7 | MFCD00016618 | 99.4% | 184.19 g/mol | C12H8O2 | 250 MG
SDP

Catalog No. 5000302844
Encompass_Preferred

Naphthalene-2,3-dicarboxaldehyde (CAS 7149-49-7) is an aromatic dialdehyde used as a derivatization reagent and research chemical. It has molecular formula C12H8O2 and molecular weight 184.19 g/mol. Reported biological activity includes ASADH inhibition (Ki ≈ 45 μM) and antifungal activity against Candida albicans (IC50 58.2 μM; MIC 12 μg/mL). Supplied as a solid for laboratory research.

  • Aromatic dialdehyde for derivatization and fluorescent labeling.
  • High purity suitable for analytical and synthetic applications.
  • Reported enzyme inhibition and antifungal activity for biological studies.
  • Compact packaging convenient for small-scale experiments.
  • Well-characterized: CAS 7149-49-7; molecular formula C12H8O2; MW 184.19 g/mol.

Catalog No. 50-003-02844 Supplier Medchemexpress LLC Supplier No. 250MGHYW016288
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naphthalene-2,3-dicarboxaldehyde (CAS 7149-49-7) is an aromatic dialdehyde used as a derivatization reagent and research chemical. It has molecular formula C12H8O2 and molecular weight 184.19 g/mol. Reported biological activity includes ASADH inhibition (Ki ≈ 45 μM) and antifungal activity against Candida albicans (IC50 58.2 μM; MIC 12 μg/mL). Supplied as a solid for laboratory research.

  • Aromatic dialdehyde for derivatization and fluorescent labeling.
  • High purity suitable for analytical and synthetic applications.
  • Reported enzyme inhibition and antifungal activity for biological studies.
  • Compact packaging convenient for small-scale experiments.
  • Well-characterized: CAS 7149-49-7; molecular formula C12H8O2; MW 184.19 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.