Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naphthalene, 2-chloro- | 91-58-7 | MFCD00035731 | 99.7% | 162.61 g/mol | C10H7Cl | 1G
SDP

Catalog No. 5000236746
Encompass_Preferred

2-Chloronaphthalene (CAS 91-58-7) is a chlorinated naphthalene used as an organic synthesis intermediate and research reagent. It is supplied as a high-purity, crystalline solid for analytical and preparative laboratory use. Quality and safety documentation, including a certificate of analysis and safety data sheet, are available from the supplier.

  • High purity: 99.7% as listed on supplier documentation.
  • Physical form: white to off-white solid, crystalline.
  • Available pack sizes: small laboratory quantities such as 1 g and 5 g.
  • Documentation: certificate of analysis and safety data sheet available.
  • Storage recommendations: store at room temperature; in solvent, store at -20°C to -80°C for extended stability.

Catalog No. 50-002-36746 Supplier Medchemexpress LLC Supplier No. HY1244511G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Chloronaphthalene (CAS 91-58-7) is a chlorinated naphthalene used as an organic synthesis intermediate and research reagent. It is supplied as a high-purity, crystalline solid for analytical and preparative laboratory use. Quality and safety documentation, including a certificate of analysis and safety data sheet, are available from the supplier.

  • High purity: 99.7% as listed on supplier documentation.
  • Physical form: white to off-white solid, crystalline.
  • Available pack sizes: small laboratory quantities such as 1 g and 5 g.
  • Documentation: certificate of analysis and safety data sheet available.
  • Storage recommendations: store at room temperature; in solvent, store at -20°C to -80°C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.