Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naphthalene, 2-ethenyl- | 827-54-3 | MFCD00004125 | 99.8% | 154.21 g/mol | C12H10 | 100 G
SDP

Catalog No. 5000302336
Encompass_Preferred

2-Vinylnaphthalene (naphthalene, 2-ethenyl-) is a vinylic aromatic reagent used as a monomer and building block in organic synthesis and polymer research. Supplied as a high-purity solid for laboratory research, the material is accompanied by analytical documentation for traceability.

  • High purity: 99.8%.
  • White to off-white solid, convenient for handling and weighing.
  • Molecular weight 154.21 g/mol; formula C12H10.
  • Recommended storage -20°C; in solution -80°C up to 6 months.
  • Certificate of analysis, safety data sheet, and datasheet available.

Catalog No. 50-003-02336 Supplier Medchemexpress LLC Supplier No. HYW015553100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Vinylnaphthalene (naphthalene, 2-ethenyl-) is a vinylic aromatic reagent used as a monomer and building block in organic synthesis and polymer research. Supplied as a high-purity solid for laboratory research, the material is accompanied by analytical documentation for traceability.

  • High purity: 99.8%.
  • White to off-white solid, convenient for handling and weighing.
  • Molecular weight 154.21 g/mol; formula C12H10.
  • Recommended storage -20°C; in solution -80°C up to 6 months.
  • Certificate of analysis, safety data sheet, and datasheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.