Learn More
Apexbio Technology LLC NAPHTHALENE-D8-50MG
SDP
Catalog No. 5000569040
Encompass_Preferred
Naphthalene-d8 (CAS No 1146-65-2) is a deuterated form of naphthalene often used in biomedical research for its stable isotopic labeling properties Originating from hydrocarbons it serves primarily in pharmacokinetic studies as a tracer due to its ability to integrate into various biological pathways without altering their function In vitro naphthalene-d8 is useful for analyzing metabolic processes often in nanomolar to low micromolar ranges It is commonly applied in cell models and animal studies to track and quantify metabolic changes improving the accuracy of drug screening and metabolic profiling Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69040 Supplier Apexbio Technology LLC Supplier No. SL102050
Naphthalene-d8 (CAS No 1146-65-2) is a deuterated form of naphthalene often used in biomedical research for its stable isotopic labeling properties Originating from hydrocarbons it serves primarily in pharmacokinetic studies as a tracer due to its ability to integrate into various biological pathways without altering their function In vitro naphthalene-d8 is useful for analyzing metabolic processes often in nanomolar to low micromolar ranges It is commonly applied in cell models and animal studies to track and quantify metabolic changes improving the accuracy of drug screening and metabolic profiling Experimental concentrations typically depend on the specific research design