Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthalene- D8, 99% D atom, MP Biomedicals™

Catalog No. ICN52039601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN52039601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN52039601 Supplier MP Biomedicals, Inc Supplier No. 0252039601

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1146-65-2
Molecular Formula C10H8
Molecular Weight (g/mol) 136.22
MDL Number MFCD00001743
InChI Key UFWIBTONFRDIAS-PGRXLJNUSA-N
Synonym naphthalene-d8, 2h8 naphthalene, perdeuteronaphthalene, naphthalene d8, perdeuterated naphthalene, 2 h? naphthalene, naphthalene-d8, analytical standard, naphthalene-d8 >98.0 atom % d, naphthalene d8 10 microg/ml in cyclohexane
PubChem CID 92148
IUPAC Name (²H₈)naphthalene
SMILES [2H]C1=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2C([2H])=C1[2H]

Specifications

Color White
Quantity 1 g
Formula Weight 136.2
Physical Form Crystalline Powder
Chemical Name or Material Naphthalene- D8, GC/MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.