Learn More
AdipoGen Naphthalenyloxy-propanediol
SDP
Supplier:  AdipoGen CDXN0073M010
Encompass_Preferred
Chemical. CAS 61248-78-0. Formula C13H14O3. MW 218.25. Synthetic Active metabolite of the anticonvulsant compound propranolol. Compound can be used as analytical reference material.
Catalog No. 50-256-0416