Learn More
Medchemexpress LLC Naphtho[1,2-b]furan, 6,7,8,9-tetrahydro-3,5,9-trimethyl-, (9S)- | 36061-18-4 | 98.0% | 214.30 | 1 MG
SDP
Catalog No. 5000365573
Encompass_Preferred
Farfugin A is a benzofuran derivative, classified specifically as a terpenoid and sesquiterpene. It originates from plants in the Asteraceae family, particularly *Ligularia japonica (Thunb.) Less.* This compound has a molecular weight of 214.30 and a chemical formula of C15H18O. It is primarily used for research purposes.
  • Derived from natural plant sources.
  • Exhibits a benzofuran core structure.
  • Belongs to the sesquiterpene class of terpenoids.
  • Available in solid form.
Catalog No. 50-003-65573 Supplier Medchemexpress LLC Supplier No. 1MGHYN15642
Farfugin A is a benzofuran derivative, classified specifically as a terpenoid and sesquiterpene. It originates from plants in the Asteraceae family, particularly *Ligularia japonica (Thunb.) Less.* This compound has a molecular weight of 214.30 and a chemical formula of C15H18O. It is primarily used for research purposes.
  • Derived from natural plant sources.
  • Exhibits a benzofuran core structure.
  • Belongs to the sesquiterpene class of terpenoids.
  • Available in solid form.