Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Naphtho[1,2-c:5,6-c']bis[1,2,5]thiadiazole-5,10-diboronic acid bis(pinacol) ester

Catalog No. 795518100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-551-8100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-551-8100MG Supplier Sigma Aldrich Supplier No. 795518100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Naphtho[1,2-c:5,6-c']bis[1,2,5]thiadiazole-5,10-diboronic acid bis(pinacol) ester

Specifications

CAS 1467776-41-5
Melting Point 377°C to 382°C
Molecular Formula C22H26B2N4O4S2
Linear Formula C22H26B2N4O4S2
Quantity 100 mg
Synonym 5,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphtho[1,2-c:5,6-c]bis[1,2,5 ]thiadiazole; PB2-NTz
Molecular Weight (g/mol) 496.22
Percent Purity 98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.