Learn More
Medchemexpress LLC Naphtho[1,2-d]thiazol-2-ylamine | 40172-65-4 | MFCD00051329 | 99.7% | 200.27 | C11H8N2S | 5 MG
SDP
Catalog No. 5000264707
Encompass_Preferred
SKA-31 (Naphtho[1,2-d]thiazol-2-ylamine, CAS 40172-65-4) is a research-grade small-molecule activator of calcium-activated potassium channels (KCa2/KCa3.1), used to probe ion channel function and endothelial or neuronal hyperpolarization in vitro and in vivo studies.
  • Potent channel activator with KCa3.1 EC50 ~260 nM and KCa2 subtype EC50s in the low micromolar range.
  • High purity for research use (≈99.7%).
  • Small molecular weight and defined composition: 200.27 g/mol; formula C11H8N2S.
  • Soluble in DMSO (≈125 mg/mL) and formulatable for in vivo dosing.
  • Supplied as a solid powder suitable for biochemical and cellular assays.
Catalog No. 50-002-64707 Supplier Medchemexpress LLC Supplier No. HY1116555MG
SKA-31 (Naphtho[1,2-d]thiazol-2-ylamine, CAS 40172-65-4) is a research-grade small-molecule activator of calcium-activated potassium channels (KCa2/KCa3.1), used to probe ion channel function and endothelial or neuronal hyperpolarization in vitro and in vivo studies.
  • Potent channel activator with KCa3.1 EC50 ~260 nM and KCa2 subtype EC50s in the low micromolar range.
  • High purity for research use (≈99.7%).
  • Small molecular weight and defined composition: 200.27 g/mol; formula C11H8N2S.
  • Soluble in DMSO (≈125 mg/mL) and formulatable for in vivo dosing.
  • Supplied as a solid powder suitable for biochemical and cellular assays.