Learn More
Apexbio Technology LLC α-Naphthoflavone,10g CAS# 604-59-1
SDP
Catalog No. 502240687
Encompass_Preferred
α-Naphthoflavone (α-NF), also known as 7,8-benzoflavone and 2-phenyl-benzo(h)chromen-4-one, is a synthetic flavone derivative. α-Naphthoflavone is a potent inhibitor of the enzyme aromatase, which implicated in converting testosterone to estrogen [1]. It can be prepared from 2-naphthol and cinnamaldehyde [2]. α-naphthoflavone functions as both an Ah receptor antagonist and an inhibitor of cytochrome P450 activity. α-naphthoflavone stimulated P450 3A4 by selectively binding and activating an otherwise inactive subpopulation of this P450 and promoting benzo[a]pyrene binding to the latter [3]. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0687 Supplier Apexbio Technology LLC Supplier No. C335310000
α-Naphthoflavone (α-NF), also known as 7,8-benzoflavone and 2-phenyl-benzo(h)chromen-4-one, is a synthetic flavone derivative. α-Naphthoflavone is a potent inhibitor of the enzyme aromatase, which implicated in converting testosterone to estrogen [1]. It can be prepared from 2-naphthol and cinnamaldehyde [2]. α-naphthoflavone functions as both an Ah receptor antagonist and an inhibitor of cytochrome P450 activity. α-naphthoflavone stimulated P450 3A4 by selectively binding and activating an otherwise inactive subpopulation of this P450 and promoting benzo[a]pyrene binding to the latter [3]. Other sizes are also available. Please inqury us for quote.