Learn More
Cayman Chemical β-Naphthoflavone, Purity: ≥98%, CAS: 6051-87-2, 1 g
SDP
Catalog No. 502949576
Encompass_Preferred
An AhR agonist; activates transcription of CYP1A1 at 1 and 10 µM in HepG2 cells; inhibits hydrogen peroxide-induced apoptosis, as well as increases the activity of CAT and SOD and decreases the activity of MDA, in SH-SY5Y cells at 10 µM; has been used as a positive control for the induction of AhR transcriptional activity
Catalog No. 50-294-9576 Supplier Cayman Chemical Supplier No. 340401 G
An AhR agonist; activates transcription of CYP1A1 at 1 and 10 µM in HepG2 cells; inhibits hydrogen peroxide-induced apoptosis, as well as increases the activity of CAT and SOD and decreases the activity of MDA, in SH-SY5Y cells at 10 µM; has been used as a positive control for the induction of AhR transcriptional activity