Learn More
Cayman Chemical NaphthofluoresceIn 50mg
SDP
Catalog No. 502953042
Encompass_Preferred
A cell-permeable inhibitor of furin (IC50 = 12 μM); inhibits furin-mediated cleavage of the pro-form of MT1-MMP, resulting in decreased levels of active MT1-MMP; inhibits the invasion of Matrigel by the human fibrosarcoma cell lines HT1080
Catalog No. 50-295-3042 Supplier Cayman Chemical Supplier No. 1305550 MG
A cell-permeable inhibitor of furin (IC50 = 12 μM); inhibits furin-mediated cleavage of the pro-form of MT1-MMP, resulting in decreased levels of active MT1-MMP; inhibits the invasion of Matrigel by the human fibrosarcoma cell lines HT1080