Learn More
Cayman Chemical Naphthofluorescein, Purity: ≥98%, CAS: 61419-02-1, 5 mg
SDP
Catalog No. 502953123
Encompass_Preferred
A cell-permeable inhibitor of furin (IC50 = 12 μM); inhibits furin-mediated cleavage of the pro-form of MT1-MMP, resulting in decreased levels of active MT1-MMP; inhibits the invasion of Matrigel by the human fibrosarcoma cell lines HT1080
Catalog No. 50-295-3123 Supplier Cayman Chemical Supplier No. 130555 MG
A cell-permeable inhibitor of furin (IC50 = 12 μM); inhibits furin-mediated cleavage of the pro-form of MT1-MMP, resulting in decreased levels of active MT1-MMP; inhibits the invasion of Matrigel by the human fibrosarcoma cell lines HT1080