Learn More
AdipoGen Naphthol AS
SDP
Catalog No. 502560357
Encompass_Preferred
Chemical. CAS 92-77-3. Formula C17H13NO2. MW BD9837. Naphthol AS 3-Hydroxy-2-naphthanilide is a fluorescent dye commonly used as a building block or intermediate to produce fluorigenic substrates, bioactive chemicals or as a coupling reagent to produce azo dyes. After cleavage of the fluorigenic substrates by esterases the free Naphthol AS can couple with diazonium salts to form different types of fluorescent azo dyes. 3-Hydroxy-2-naphthanilide 3HNA is also a naturally occurring chemical that can be found in human urine. It is used as a marker for early pregnancy.
Catalog No. 50-256-0357 Supplier AdipoGen Supplier No. CDXN0529G025
Chemical. CAS 92-77-3. Formula C17H13NO2. MW BD9837. Naphthol AS 3-Hydroxy-2-naphthanilide is a fluorescent dye commonly used as a building block or intermediate to produce fluorigenic substrates, bioactive chemicals or as a coupling reagent to produce azo dyes. After cleavage of the fluorigenic substrates by esterases the free Naphthol AS can couple with diazonium salts to form different types of fluorescent azo dyes. 3-Hydroxy-2-naphthanilide 3HNA is also a naturally occurring chemical that can be found in human urine. It is used as a marker for early pregnancy.