Learn More
Medchemexpress LLC Naphthol AS-BI | 1237-75-8 | 99.86% | 250 MG
SDP
Catalog No. 5000413733
Encompass_Preferred
Naphthol AS-BI is a substrate of β-glucuronidase and produces a bright red effect after staining biological tissues. It is intended for research use only.
  • Substrate for β-glucuronidase
  • Produces bright red effect
  • Suitable for staining biological tissues
  • For research use only
Catalog No. 50-004-13733 Supplier Medchemexpress LLC Supplier No. 250MGHYD0164
Naphthol AS-BI is a substrate of β-glucuronidase and produces a bright red effect after staining biological tissues. It is intended for research use only.
  • Substrate for β-glucuronidase
  • Produces bright red effect
  • Suitable for staining biological tissues
  • For research use only