Learn More
Medchemexpress LLC Naphthol AS-BI | 1237-75-8 | 99.86% | 50 MG
SDP
Catalog No. 5000410521
Encompass_Preferred
Naphthol AS-BI serves as a substrate for β-glucuronidase, enabling a bright red staining effect in biological tissues. This product is intended for research applications.
  • Acts as a substrate for β-glucuronidase
  • Produces a bright red staining effect
  • Suitable for biological tissue staining
  • Designed for research use
  • Presented as a solid
  • Chemically stable under recommended storage conditions
Catalog No. 50-004-10521 Supplier Medchemexpress LLC Supplier No. HYD016450MG
Naphthol AS-BI serves as a substrate for β-glucuronidase, enabling a bright red staining effect in biological tissues. This product is intended for research applications.
  • Acts as a substrate for β-glucuronidase
  • Produces a bright red staining effect
  • Suitable for biological tissue staining
  • Designed for research use
  • Presented as a solid
  • Chemically stable under recommended storage conditions